VWR® Sample Vials, with Snap-Caps
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
216-1849EA 1270 DKK
216-1849 216-1848 216-1847 216-1851 216-1846 216-1845 216-1844 216-1843
VWR® Sample Vials, with Snap-Caps
Hætteglas
Soda-lime glass, amber.
  • Ideal for storing medication as liquid, powder or capsules
  • Used for light-sensitive liquids
  • White snap-caps are impervious to ultraviolet light
  • Not suitable for heat applications
Leveringsinformation: Delivered in practical safe trays. PE snap-caps are included.