Derivatisation reagent, phenylisothiocyanate, Pierce™
Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Pierce
PIER26922EA 1300 DKK
PIER26922
Derivatisation reagent, phenylisothiocyanate, Pierce™
Kromatografi sorbenter HPLC Sorbenter
Phenylisothiocyanate (PITC) is a high-purity reagent for pre-column quantitative derivatisation of amino acids by reverse-phase HPLC.
  • Produces stable products with all amino acids, including proline
  • For pre-column derivatisation of amino acids for picomole detection levels in HPLC
  • Produces phenylthiocarbamyl (PTC) derivatives that can be separated and quantified using reverse HPLC
Phenylisothiocyanate (PITC), also known as Edman's Reagent, enables the sequential degradation of amino acids in a polypeptide chain, yielding primary structural information. PITC reacts readily with amino acids at alkaline pH. Precolumn derivatisation results in phenylthiocarbamyl derivatives. Reacts with amine groups of amino acids in 5 to 10 minutes at room temperature to yield products that are detectable by UV absorbance (λmax = 254 nm)