Serum replacement, HyClone AdvanceSTEM™
Leverandør: HyClone products (Cytiva)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HYCLSH30874.02EA 638 DKK
HYCLSH30874.02 HYCLSH30874.03
Serum replacement, HyClone AdvanceSTEM™
HyClone™ AdvanceSTEM™ Serum Replacement has been developed to support the growth of undifferentiated pluripotent embryonic stem cells in culture with no requirement for foetal bovine serum (FBS) supplementation. The product is intended for use with HyClone™ AdvanceSTEM™ Low Osmo DMEM.
  • No requirement for foetal bovine serum
  • Designed to maintain mESCs in an undifferentiated state
  • Not recommended for plating mouse embryonic feeder cells (MEFs)