Digital anemometer/thermometer, LU8000
Leverandør: Industrial Physics
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SIMELU8000EA 2060 DKK
SIMELU8000
Digital anemometer/thermometer, LU8000
Digital wheel anemometer with a built-in thermometer for measuring air velocity in for example ventilation ducts, air conditioning, etc.
  • 'Hold' function for holding the measured values
  • 4-digit LCD display with backlight
  • Custom LSI microproessor circuit Dual function 0,7" (16 mm)
  • Air velocity/flow sensor: Conventional angled vane arms with low-friction bali bearing
  • Temperature sensor: NTC type
  • Auto shut oft after 20 minutes
Leveringsinformation: Supplied with probe and battery.