2-(4-Phenoxyphenyl)ethylboronic acid diethanolamine ester 97%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

L19703.MCEA 491 DKK
L19703.MC
2-(4-Phenoxyphenyl)ethylboronic acid diethanolamine ester 97%
2-(4-Phenoxyphenyl)ethylboronic acid diethanolamine ester
Formel: C₁₈H₂₂BNO₃
Smeltepunkt: 178…191 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD12546199
CAS-nummer: 608534-43-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR