Substance P

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

J63489.MBEA 7570 DKK
J63489.MB
Substance P
Proteiner
CAS No.: 33507-63-0
Molecular Formula: C63H98N18O13S
Formula Weight: 1347,64
Physical Form: Powder
Appearance: White
MDL No.: MFCD00076780

Applications: A neurokinin NK-1 receptor agonist
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR