RNA sample loading buffer (6X)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
J62468.XFEA 400 DKK
ALFAJ62468.XF ALFAJ62468.S0
RNA sample loading buffer (6X)
Storage Temperature: −30 to −10 °C
Physical Form: Liquid
MDL No.: MFCD00213041