2-Bromo-3,5-dichloroisonicotinic acid 95%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H63739.MDEA 366 DKK
H63739.MD H63739.06 H63739.03
2-Bromo-3,5-dichloroisonicotinic acid 95%
2-Bromo-3,5-dichloroisonicotinic acid
Formel: C₆H₂BrCl₂NO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03411637
CAS-nummer: 343781-56-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR