N-Boc-2-cyclopentyl-L-glycine dicyclohexylammonium salt 95%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H62838.MDEA 445 DKK
H62838.MD
N-Boc-2-cyclopentyl-L-glycine dicyclohexylammonium salt 95%
N-Boc-2-cyclopentyl-L-glycine dicyclohexylammonium salt
Formel: C₁₂H₂₁NO₄·C₁₂H₂₃N
MDL Number: MFCD05663437
CAS-nummer: 676559-50-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR