5-Brom-2-chlornicotinsyre 96%
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H61154.22EA
2990
DKK
H61154.22
5-Brom-2-chlornicotinsyre 96%
5-Brom-2-chlornicotinsyre
Formel:
C₆H₃BrClNO₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD03844847 CAS-nummer: 29241-65-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...