2-Amino-3-benzyloxy-5-bromopyridine 96%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H54667.03EA 2390 DKK
H54667.03 H54667.MD
2-Amino-3-benzyloxy-5-bromopyridine 96%
2-Amino-3-benzyloxy-5-bromopyridine
Formel: C₁₂H₁₁BrN₂O
Molekylvægt: 279,13
Smeltepunkt: 92…94 °C
MDL Number: MFCD11109946
CAS-nummer: 754230-78-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR