2-Amino-3,7-dihydropyrrolo[2,3-d]pyrimidin-4-one 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H54595.MDEA 891 DKK
H54595.MD
2-Amino-3,7-dihydropyrrolo[2,3-d]pyrimidin-4-one 97%
2-Amino-3,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
Formel: C₆H₆N₄O
Molekylvægt: 150,14 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09033259
CAS-nummer: 7355-55-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR