4-Ethoxy-2-fluorobenzoyl chloride 97%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

H33851.06EA 6000 DKK
H33851.06 H33851.03
4-Ethoxy-2-fluorobenzoyl chloride 97%
4-Ethoxy-2-fluorobenzoyl chloride
Formel: C₉H₈ClFO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13196594
ADR: 8,II

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR