7-Fluorochroman-4-one 98% may contain up to 15% 5-isomer (sum of isomers)
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H31881.MDEA
641
DKK
H31881.MD
H31881.03
7-Fluorochroman-4-one 98% may contain up to 15% 5-isomer (sum of isomers)
7-Fluorochroman-4-one
Formel:
C₉H₇FO₂ Molekylvægt: 166,15 g/mol Smeltepunkt: 46…50 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD06202344 CAS-nummer: 113209-68-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...