Benzyl(4-oxocyclohexyl)carbamate 97%
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H26015.06EA
3160
DKK
H26015.06
Benzyl(4-oxocyclohexyl)carbamate 97%
Benzyl(4-oxocyclohexyl)carbamate
Formel:
C₁₄H₁₇NO₃ Molekylvægt: 247,29 g/mol Smeltepunkt: 81…83 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD06658393 CAS-nummer: 16801-63-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...