Diethyl dimethylaminomethylenemalonate 95%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B20765.06EA 1060 DKK
B20765.06 B20765.14
Diethyl dimethylaminomethylenemalonate 95%
Diethyl dimethylaminomethylenemalonate
Formel: (CH₃)₂NCH=C(CO₂CH₂CH₃)₂
Molekylvægt: 215,25 g/mol
Kogepunkt: 134…136 °C (0,5 mmHg)
MDL Number: MFCD00052394
CAS-nummer: 18856-68-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR