VWR® Toronto, Safety Goggles
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR® Toronto
111-1840EA 118 DKK
111-1840
VWR® Toronto, Safety Goggles
Beskyttelsesbriller
Wrap-around modern style provides excellent panoramic vision, high level of comfort and maximum protection.
  • PC lens, PVC frame, polyester/rubber headband
  • Fitted with indirect ventilation
  • Opalescent frame prevents light reflection
  • Designed to give optimum fit to all facial contours
  • Comfortably fits over most spectacles
Certificeringer: Compliant with: EN 166:2001. Frame: EN166 3 4 9 BT CE. Lens: VWR® 1 BT 9.