Protective hand and body cream, Stokoderm® Advanced
Leverandør: SC Johnson
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EVON34770EA 2650 DKK
EVON34770
Protective hand and body cream, Stokoderm® Advanced
Universal pre-work cream with natural film building agents to provide advanced protection. It helps to protect the skin against water and non water-based workplace contaminants, for example, oils and solvents in areas with heavy contamination or changing workplace substances. Suitable for use on hands and body. For areas were gloves are not worn.
  • Universal pre-work cream with natural film building agents to provide advanced protection
  • Contains unique oil-suspending emulsifiers to make skin easier to clean
  • Effective against concentrated water and heavy non water-based workplace contaminants
  • Colourant- and silicone-free