VWR®, Powder sampler
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
300-0166EA 1600 DKK
300-0166
VWR®, Powder sampler
Prøvetagere Prøveøser
Sampler probe specially designed for single samples from PE or paper bags. The needle point sampler punctures the bag with minimum damage.
Suitable for use in the pharmaceutical, chemical, also food and dairy industries for collecting powder, flour, crystals and sugar.
Method of use: The probe is pulled out with sample ready to be poured out of the hollow handle. A bag or a container can be positioned over the handle for sample collection. The hole on the side of the bag can be closed with adhesive tape or with an appropriate label.