Ice Makers CoolNat Propane (R290)
Leverandør: Ziegra Eismaschinen
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Coolnat Propane (R290)
474-0169EA 61140 DKK
474-0169 474-0147 474-0148 474-0170 474-0171 474-0172 474-0150 474-0173 474-0151 474-0174 474-0152 474-0154 474-0155 474-0156 474-0157 474-0158 474-0159 474-0160 474-0161 474-0140 474-0162 474-0163 474-0141 474-0142 474-0164 474-0143 474-0165 474-0144 474-0166 474-0145 474-0167 474-0146 474-0168
Ice Makers CoolNat Propane (R290)
Ismaskiner
Ice machines for small and moderate requirements with storage and other accessories, this extensive range of propane (R290) ice machines offers solutions for all applications. Ice machines in the CoolNat propane series come equipped with high quality safety valves and the most up-to-date pressure monitors to ensure the safest possible use of propane. Minimal amounts of refrigerant are used, well within the levels set by the most stringent of technical safety requirements. The CoolNat range offers the same features than the standard serie.
  • Choice of micro or standard ice sizes ensure the most appropriate kind of ice for any application
  • As with all ZIEGRA ice machines, those in the CoolNat series are also designed to be especially service-friendly
  • Ready to go: simply connect to power and a potable or demineralised water supply
  • Easy to store: the ice is always loose and freeflowing
  • Air or water-cooled options on all R290 machines
Cabinet and bin made of stainless steel.
Ice storage bin must be ordered separately.
Bestillingsoplysninger: Three models - ZBE 70, 150 and 250 do not have integrated bins. These need to be ordered separately. See ordering table below for details.
Leveringsinformation: All installation fittings included.