Human recombinant MANF
Leverandør: Tonbo Biosciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
21-7111-U025EA 1370 DKK
TNBO21-7111-U025 TNBO21-7111-U100 TNBO21-7111-U500 TNBO21-7111-U005
Human recombinant MANF
Mesencephalic astrocyte-derived neurotrophic factor (MANF), also known as ARMET or ARP, is a secreted protein belonging to the ARMET family. It is localised to the Golgi apparatus and the endoplasmic reticulum (ER), and has been reported to play a role in modulating myocardial damage from ER stress and has been called a cardiomyokine. MANF, like its structural homolog CDNF, has been described as a survival factor for midbrain dopaminergic neurons in vitro and in vivo.