Human recombinant DKK3
Leverandør: Tonbo Biosciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
21-7102-U010EA 1400 DKK
TNBO21-7102-U010 TNBO21-7102-U002 TNBO21-7102-U100 TNBO21-7102-U500
Human recombinant DKK3
The DKK (Dickkopf) family of secreted proteins includes DKK-1 through -4, and a related protein, Soggy (DKK-11). DKK-3 exhibits the most sequence homology with Soggy. It is expressed in many normal tissues including brain, spinal cord and heart. Also known as REIC (Reduced Expansion in Immortalised Cells), DKK-3 may be a negative growth regulator as expression is downregulated in a variety of cancer cell lines and tumors. Unlike other DKK members, it has been suggested that DKK-3 does not bind to LRP5 and/or LRP6.