Human recombinant CDNF
Leverandør: Tonbo Biosciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
21-7099-U025EA 1370 DKK
TNBO21-7099-U025 TNBO21-7099-U100 TNBO21-7099-U500 TNBO21-7099-U005
Human recombinant CDNF
Cerebral Dopamine Neurotrophic Factor (CDNF), also known as ARMETL1, is a novel secreted protein that exhibits strong trophic activity on dopaminergic neurons, comparable to that of GDNF. It is structurally similar to MANF. This conserved neurotrophic factor promotes the survival, growth and function of dopaminergic neurons. Besides neurons, it is widely expressed in brain regions including cerebral cortex, cerebellum and hippocampus.