Mouse recombinant CCL4
Leverandør: Tonbo Biosciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
21-7098-U500EA 13990 DKK
TNBO21-7098-U500 TNBO21-7098-U010 TNBO21-7098-U002 TNBO21-7098-U100
Mouse recombinant CCL4
CCL4 (MIP-1β) is a mitogen-inducible monokine and member of the CC chemokine family. It is a secreted protein that plays a role in the acute inflammatory response and is a chemoattractant for monocytes, neutrophils, eosinophils, NK cells, and CD4+ T lymphocytes. MIP-1β can signal through CCR5 and, by inducing downregulation of this receptor, can block HIV-1 infection of T cells.