Liquid nitrogen transport carrier, CryoPod™
Leverandør: BioCision
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BCISBCS-514EA 31360 DKK
BCISBCS-514
Liquid nitrogen transport carrier, CryoPod™
CryoPod™ Carrier provides safe, reliable and portable <-150 ˚C cryogenic transport for biospecimens for over 4 hours. Use around the laboratory, campus or short transport between facilities. Displays temperature, date and time, and features audible and visual alarms, as well as logging capability.
  • No direct sample contact with LN₂
  • With built-in handle and finger grips
  • Magnetised foam lid for safer transport and insulation
  • Accomodates one standard cryogenic storage box or 2 to 3 small cassettes
It integrates into an (optional) automated filling station or can be manually filled using the included fill kit.
Bestillingsoplysninger: CryoPod™ Carrier includes green lid and manual fill kit. Optional CryoPod lids available in 4 additional colours.