Temperature chart recorders, KT
Leverandør: Dickson
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
DICKKT6P1EA 5140 DKK
DICKKT6P1 DICKKT6P2 DICKKT6P5
Temperature chart recorders, KT
This line of temperature chart recorders feature a sleek look and feel, precision pen movement, k-thermocouple probes and 8 user selectable temperature ranges to choose from. The 6" (152 mm) is a great size for monitoring refrigerators and freezers.
  • One K-Thermocouple probe
  • 6" chart (152 mm)
  • User selectable recording times and 8 temperature ranges
  • AC power with battery backup
  • Replaceable sensors
  • IP rating: 21
  • Enclosure: Rugged black ABS case, polycarbonate door, aluminium dial
Certificeringer: CE and RoHs approvals
Leveringsinformation: AC Adapter, red Pen, 9 V battery, quick start guide and 4' bead-wire probe are included. Charts must be purchased separately.