Ammeter, analogue, multi range
Leverandør: Eisco Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EISCPH1108BEA 1710 DKK
EISCPH1108B
Ammeter, analogue, multi range
The operation of the meter depends on the reaction of the current circulating in the moving coil and a field of a fixed permanent magnet.
  • AC and DC current
  • Multi range: 0 to 10 mA, 100 mA, 300 mA, 1 A, 3 A, 5 A
  • Linear scale