Voltmeter, analogue, multi range
Leverandør: Eisco Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EISCPH1108AEA 1710 DKK
EISCPH1108A
Voltmeter, analogue, multi range
The operation of the meter depends on the reaction of the current circulating in the moving coil and a field of a fixed permanent magnet.
  • AC and DC voltages
  • Multi range: 0 to 1 to 10 to 30 to 100 to 300 to 500 Volts
  • Linear scale