Bump cap, First Base 3 Elite
Leverandør: Scott Safety
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SCTS2021047EA 162 DKK
SCTS2021047
Bump cap, First Base 3 Elite
This stylish bump cap is made from lightweight, microfibre fabric. Its unique shell design delivers optimum protection and is shaped over the ear to provide enhanced compatibility with other forms of PPE.
  • Microfibre fabric reduces humidity and allows air circulation
  • Side mesh ventilation enhances breathability
  • Uses a unique flexible shell design to provide an optimum fit
  • Elasticated adjustment strap positions beneath the occipital bone for increased stability