Full face masks, FM4
Leverandør: Scott Safety
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SCTS2024413EA 2310 DKK
SCTS2024413
Full face masks, FM4
FM4 is a full facemask made from silicone rubber providing the highest levels of respiratory protection. A novel hose connection brings the hose up over the wearer’s shoulder, keeping the work area clear. The mask has an ultra comfortable silicone face seal and offers a virtually unrestricted field of vision.
  • Ultimate comfort with LSR silicone seal and unrestricted (98%) field of vision
  • Side entry hose improves comfort and balance
  • Full facemask available in three sizes, also approved for unpowered use with Pro2000 filters
  • Compatible blower units: Spirit, Tornado, Proflow2 120SC / 120EX, 160SC / 160EX and 120EX Proflow3
Certificeringer: EN 12942 TM3, AS/NZS1716-P3
Bestillingsoplysninger: FM4 is supplied without hose, to be ordered separately.