Digital multimeter, autoranging with non contact voltage detector, MS8360F
Leverandør: EDUCATIONSCIENCE COLLECTION P
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EDSCMS8360FEA 0 DKK
EDSCMS8360F
Digital multimeter, autoranging with non contact voltage detector, MS8360F
Multimetre Digitale multimetre
Multimeter to test DC voltage, AC voltage, DC current, AC current, resistance, capacitance, frequency and duty cycle.