Half mask respirator, FM1
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SCTS2023539EA 591 DKK
SCTS2023539 SCTS2023538
Half mask respirator, FM1
The reusable half mask FM1 is designed with liquid silicone rubber face seal for exceptional comfort and fit. The low profile design of the mask makes it compatible with visors, welding shields and other PPE. Ideal for welding and spraying.
  • Integrates easily with other PPE
  • Side entry hose improves comfort and balance
  • Compatible with Spirit, Tornado, Proflow2 120SC/120EX, 160SC/160EX & Proflow3 120EX
Certificeringer: EN12942 TM2
Bestillingsoplysninger: FM1 is supplied without hose and that should be ordered separately.