Visors for safety helmets Style 300, Style 600, Tuffmaster II
Leverandør: Scott Safety
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SCTS2019456EA 1770 DKK
SCTS2019456 SCTS2017866
Visors for safety helmets Style 300, Style 600, Tuffmaster II
The Protector FHK67 Electrician’s helmet-mounted visor carrier range is suitable for use as protection against short circuit electric arc of up to 1000 V AC. The visor consists of an acetate visor and a PVC visor carrier.
  • Easy attachment to the helmet via a rubber band
  • No metal components
  • Does not interfere with wearing hearing protection
  • Compatible with Style 300, Style 600, Tuffmaster II safety helmets
Colour: Clear
Certificeringer: EN 166 2 1.2 2B38, VDE, GSET29: Class 1