Speed of sound kit
Leverandør: EDUCATIONSCIENCE COLLECTION P
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EDSC241-513EA 4540 DKK
EDSC241-513
Speed of sound kit
Undervisningsmateriale Fysik undervisningsmateriale Waves and Oscillations Learning Activities
The Speed of Sound Kit is extremely simple to set up and use, and provides accurate and precise measurements for the speed of sound in air. The Edulab Speed of Sound Kit comes with two condenser microphones (including leads), to be placed a metre apart and used as start-stop signals in detecting the passing sound wave, and suitable tools for generating the correct type of sound. Also included is a microsecond timer which can be used as a start-stop microsecond or millisecond timer, for use with light gates or mechanical switches in a wide range of other experiments.