The natural display (Natural-V.P.A.M.) shows mathematical expressions like roots and fractions as they appear in your textbook, and this increases comprehension because results are easier to understand.
- 252 functions
- Natural textbook display
- Two-way power
Functions: New fraction input mode - Random integers - Prime factorization - Repeat function - 24 levels of brackets - Variable memories (6) - STO/RCL keys - Scientific functions: Trigonometric and inverse trigonometric functions - Hyperbolic and inverse hyperbolic functions - Exponential functions 10x, ex - Inverse function n x-1 - Power and root functions x√, √, x2, xy - Logarithm functions (log, ln) - Combinatoric and permutations - Random number generator - Converts between polar coordinates and rectangular coordinates - Calculates with fractions (two modes) - Converts between sexagesimal and decimal - Calculates in gradian, degree and radian measure - π key, e key und % key - SCI/FIX - Statistics.
Leveringsinformation: Supplied with a hard case.