Bump cap, First Base 3 Flame Retardant
Leverandør: Scott Safety
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
First Base 3 Flame Retardant
SCTS2021254EA 258 DKK
SCTS2021254
Bump cap, First Base 3 Flame Retardant
The First Base 3 flame-retardant bump protection cap is designed to protect workers where they are exposed to flammable liquids or hot sparks that could potentially burn the head or ignite hair.
  • No direct ventilation holes minimises potential burn risk
  • Completely flame-retardant (coated cotton) outer cap
  • High performance sweatband absorbs and wicks moisture away quickly
  • Flexible ABS protective shell guards against bumps and lacerations to the head
Colour: Navy blue
Certificeringer: EN 812:A1