Transfer trolley Mielcar
Leverandør: Miele
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
141-0520EA 0 DKK
141-0520
Transfer trolley Mielcar
Vogne
For loading and unloading washer-disinfectors with backets and inserts.
  • 4 castors (2 are lockable)
  • Height adjustable
  • Two recessed shelves, handle and docking system