Transport trolley, MT Mieltrans
Leverandør: Miele
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
141-0393EA 0 DKK
141-0393
Transport trolley, MT Mieltrans
Vogne
For storing and transporting baskets and inserts on 4 levels.
  • Adjustable by up 102,5 mm
  • Running on 4 lockable castors