Digital stopwatch/repeat timer, Traceable®, VWR®
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWRU62374-000
Digital stopwatch/repeat timer, Traceable®, VWR®
Digital stopwatch with shockproof ABS plastic casing.
  • Timing capacity: 9 hours, 59 minutes, 59 seconds and 99 hundredths
  • Functions: Single action, time-in/-out, cumulative split, interval split, continuous timing
  • Dual display shows cumulative and interval split
  • Repeat timer provides repeat function of countdown alarm timing
  • When zero is reached, timer alarms and repeats countdown from the programmed time
  • Eight recallable memories
  • Clock function
Certificeringer: Compliance with ISO/IEC 17025; accredited by A2LA indicating traceability to standards provided by NIST.
Leveringsinformation: Supplied with lanyard, battery and Traceable® certificate.