VWR®, Digital Stopwatch
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
609-0214EA 593 DKK
609-0214
VWR®, Digital Stopwatch
Alarmure Digitale stopure
Quartz-crystal stopwatch with dual display.
  • Timing capacity: 9 hours, 59 minutes, 59 seconds and 99 hundredths
  • Functions: Single action, time out, cumulative split, interval split and continuous roll-over timing
  • Dual display shows cumulative and interval split
  • Repeat timer provides repeat function of countdown alarm timing
  • Eight recallable memories, plus fastest and slowest lap times
  • Tough, water resistant ABS plastic casing
  • Ideal for enzyme tests, viscosity measurements and all timed lab tests
Certificeringer: Compliance with ISO/IEC 17025; accredited by A2LA indicating traceability to standards provided by NIST.
Leveringsinformation: Supplied with lanyard, battery and certificate.