Vil du logge ind igen?
OBS! Vi har skiftet mail domæne til @vwr.com så det er ikke længere muligt at skrive til @dk.vwr.com.
Anti-PHOX2B Rabbit Polyclonal Antibody
Leverandør: BioVision
This gene encodes a member of the bicoid sub-family of homeodomain-containing transcription factors. The encoded protein acts as a transcription factor and may play a role in brain and sensory organ development. A similar protein in mice is required for proper brain and sensory organ development and can cause epilepsy.
Recommended Dilutions: ELISA: 1:1,000; Western Blot: 1: 100~500).
Type: Primary
Antigen: PHOX2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
BESTIL
Udvid alle / Skjul alle
Størrelse Pakn Pack type VWR katalog nummer Mængde
100 µg 100 µG Vial BIOV3919-100
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Specifikationer
Antigen symbol PHOX2B
Antigen name Paired-like homeobox 2B
Reactivity Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
Antigen synonyms PMX2B, NBLST2, NBPhox
Gene ID 8929
Koncentration 0,25 mg/ml
Cross adsorption No
Immunogen KLH conjugated synthetic peptide between 111-138 amino acids from the Central region of human PHOX2B.
Purification Affinity purified
Form Liquid
Storage buffer Supplied in PBS with 0,09% (W/V) sodium azide at a concentration of 0,25 mg/ml. This antibody is purified thro?gh a protein A column, followed by peptide affinity purification.
Opbevaringstemperatur – 20 °C
Shipping temperature 4 °C
Skriv en bedømmelse
Brug for hjælp?
  • Uanset hvad du mener, positivt eller negativt, er det vigtigt for os.
  • Det tager kun få sekunder at give os feedback.
  • Din feedback vil blive offentliggjort på internettet.
VWR.com vil offentliggøre dit navn som vwr.com bruger.
Du kan bruge dette navn, eller redigere og gemme et kaldenavn.
Vi anbefaler at brugerne bruger deres første og sidste initialer
Bekræft venligst branchen du arbejder i
Eksempel: farmaceutisk
Bekræft venligst dit arbejde
Eksempel: Klinisk assistent
Bekræft venligst Byen
Eksempel: New York
Bedømmelser
Samlet vurdering
Gennemsntilig vurdering
Sortér alle efter:
Enter Details
Select the article for which you are looking for Certificate
Anfør Lotnummeret. Feltet er mandatory. skriv "x" for VWR kemikalier hvis du ikke kender nummeret
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse katalog Nummer Availability Enhed Din pris Price Per Antal