Keep my session open?
Ending In 
The session is expired
Din session er udløbet. For din sikkerhed har vi logget dig ud.
Vil du logge ind igen?

Anti-ANGPT1 Rabbit Polyclonal Antibody

Leverandør: BioVision
Angiopoietin-1 (Ang-1) is a secreted ligand for Tie-2, a tyrosine-kinase receptor expressed primarily on vascular endothelial cells and early hematopoietic cells. Ang-1/ Tie-2 signaling promotes angiogenesis during the development, remodeling, and repair of the vascular system. Transgenic mice lacking expression of either Ang-1 or Tie-2 fail to develop a fully functional cardiovascular system and die before birth. Postnatally, the angiogenic activity of Ang-1/Tie-2 is required during normal tissue repair and remodeling of the female endometrium in the menstrual cycle. Ang-1/Tie-2 signaling appears to be regulated by Angiopoietin-2 (Ang-2), a natural antagonist for Tie-2 that exerts its effects through an internal autocrine loop mechanism. In addition to suppressing endothelial cell activation by inhibiting the expression of adhesion and inflammatory molecules, Ang-1 enhances endothelial cell survival and capillary morphogenesis, and lessens capillary permeability. As such, Ang-1 has a potential to become an effective therapeutic agent for treating various endothelium disorders, including several severe human pulmonary diseases. The efficacy of cell-based Ang-1 gene therapy for acute lung injury (ALI) has recently been studied in a rat model of ALI. The results of this study show that such therapy can markedly improve lung condition and suggest that Ang-1 therapy may represent a potential new strategy for the treatment and/or prevention of acute respiratory distress injury (ARDI), a significant cause of morbidity and mortality in critically ill patients. Recombinant human ANG-1, derived from HeLa cells, is a C-terminal histidine tagged glycoprotein which migrates with an apparent molecular mass of 60.0 – 70.0 kDa by SDS-PAGE under reducing conditions. Sequencing analysis shows N-terminal sequences starting with Ser-20 and with Asp-70 of the 498 amino acid precursor protein.

Recommended Dilutions: Western Blot: ~1:1000.

Type: Primary
Antigen: ANGPT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human. Predicted Cross Reactivity With Mouse, Bovine And Pig Samples.



 

BESTIL

Udvid alle / Skjul alle
Størrelse Pakn Pack type VWR katalog nummer Mængde
100 µl 100 µl Vial BIOV6760-100
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Specifikationer

Antigen symbol ANGPT1
Antigen name angiopoietin 1
Reactivity Pig, Mouse, Bovine, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
Antigen synonyms AGP1, ANG1, AGPT
Gene ID 284
Epitope C-terminal
Cross adsorption No
Immunogen This ANGPT1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 374-402 amino acids from the C-terminal region of human ANGPT1.
Purification This antibody is purified through a protein A column, followed by peptide affinity purification.
Form Liquid
Storage buffer PBS with 0,09% (W/V) sodium azide
Opbevaringstemperatur – 20 °C
Shipping temperature 4 °C

Skriv en bedømmelse

Brug for hjælp?

  • Uanset hvad du mener, positivt eller negativt, er det vigtigt for os.
  • Det tager kun få sekunder at give os feedback.
  • Din feedback vil blive offentliggjort på internettet.

VWR.com vil offentliggøre dit navn som vwr.com bruger.
Du kan bruge dette navn, eller redigere og gemme et kaldenavn.
Vi anbefaler at brugerne bruger deres første og sidste initialer

Bekræft venligst branchen du arbejder i

Eksempel: farmaceutisk

Bekræft venligst dit arbejde

Eksempel: Klinisk assistent

Bekræft venligst Byen

Eksempel: New York

Bedømmelser

Samlet vurdering
Gennemsntilig vurdering

Sortér alle efter:

Enter Details

Select the article for which you are looking for Certificate

Anfør Lotnummeret. Feltet er mandatory. skriv "x" for VWR kemikalier hvis du ikke kender nummeret

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivelse katalog Nummer Availability Enhed Din pris Price Per Antal
 

Et nyt udseende og et fornyet engagement

Bygget på de funktioner og ressourcer, du kender, og optimeret med innovation fra Avantor®. Vores nye brand symboliserer agilitet og vores evne til at tilpasse os kundernes behov.

LÆS MERE BEGYND DIN SHOPPING