CO₂ incubators with hot air sterilisation, C Series,
Leverandør: Binder
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
390-0925EA 59170 DKK
BIND9040-0112
CO₂ incubators with hot air sterilisation, C Series,
The C-170 CO₂ incubator provides ideal, reproducible growth conditions for cell cultures. The unique deep-drawn stainless steel, obstruction free chamber makes cleaning very easy. Uniform growth conditions are provided by the VENTAIR™ jacket system, this ensures accurate temperatures, homogeneous temperature distribution and enables fast recovery times.
  • Seamless, deep-drawn inner stainless steel chamber without fan and a two-part rack without screw connections for increased hygiene
  • Auto-sterilisation with hot air at 180 °C eliminates spores and bacteria
  • Controller with LED display for simultaneous display of temperature, and CO₂ concentration
  • CO₂ introduction through Venturi gas mixing nozzle
  • Electronic error self-diagnostic system with visual and acoustic alarms, as well as potential free contact for central monitoring
  • Independent temperature safety device Class 3.1 (DIN 12880) with visual and audible temperature alarm, which protects the samples and the incubator
  • Stable pH values due to short CO₂ recovery times with drift-free infrared CO₂ measurement system
It also minimises thermal degradation of the cell growth due to occasional door openings and the Permadry™ double pan humidication system ensures high humidity and condensate-free interior walls. Units can be stacked using the stacking adaptor to save space, have a tightly-fitted inner glass door made of safety glass and 2-point door lock.
Leveringsinformation: Supplied with 3 perforated stainless steel shelves and Binder test certificate. Optional ethernet interface for APT-COM™ DataControlSystem communication software. Calibrations and validations possible, for details please contact your local VWR sales office.