VWR® INCU-Line® Prime, Incubators with Natural Convection
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
390-0910EA 13730 DKK
390-0910 390-0911
VWR® INCU-Line® Prime, Incubators with Natural Convection
Inkubatorer Standardinkubatorer
These incubators with natural air convection feature a stainless steel inner chamber, a powder coated sheet exterior and an internal glass door. All models have a 30 mm Ø access port, real time clock, digital timer, USB port and test results memory function to recall temperature data or events with an internal memory which stores up to 2046 data records. The range has an auto-diagnostic function, temperature sensor fail alarm, door lock, open door alarm and audible and visual temperature alarms. All models have the following safety features: Independent adjustable over-temperature safety device Class 3,1 protects the samples and the incubator and a power failure control system (program continues after power restored). All Prime oven and incubator units of the same size can be stacked without additional accessories (except the 180 L models which are not stackable).
  • Microprocessor controller with LCD graphic display and illuminated touch buttons
  • Stores up to 3 programs, each program includes max. 3 adjustable ramps and 3 temperature-time profiles
  • Loop function up to 99 times or endless
  • Adjustable hold at set point time for temperature from 1 min to 5 days or continuous operation
  • Manual air flap control, 0% (closed), 25%, 50%, 75%, 100% (open)
Leveringsinformation: Supplied with 2 stainless steel wire shelves for 56 and 112 Prime models and 3 shelves for the 180 Prime model. Manufacturer test certificate (data measured at 37 °C) and Basic Control Software to download data from internal memory. Multilingual instruction manual.