Laboratory tables, polymer, MetroMax i®
Leverandør: Metro International
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
METOLT30XPB3EA 2750 DKK
METOLT30XPB3
Laboratory tables, polymer, MetroMax i®
These polymer laboratory workbenches available with black phenolic tops and solid metromax i shelves constructed of polymer with top phenolic resin.
  • The top has good abrasion, impact resistance and stain resistance
  • The posts are all polymer and accept all MetroMax iQ casters
  • The bottom shelf is a solid MetroMax i polymer shelf with built-in Microban antimicrobial protection
  • Maximum load capacity is upto 273 kg
* Mobile posts include an adaptor insert that will accept Metro 127 mm stem casters.