Adhesives, DP 8005, Scotch-Weld™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MMMA8005T265EA 2130 DKK
MMMA8005T265
Adhesives, DP 8005, Scotch-Weld™
Scotch-Weld™ Structural plastic adhesive DP8005 is a two-part acrylic based adhesive (10:1 ratio by volume) that can bond many low surface energy plastics, including many grades of PP, PE and TPE (thermoplastic elastomers) without special surface preparation.
  • Solvent-free adhesive system
  • Excellent water and humidity resistance
  • Excellent chemical resistance
  • Component materials: Acrylate
  • Fixture time: 30 min
  • Viscosity: 7 mPa
  • Colour: White