Electrode stand, E190
Leverandør: Hach
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HACH809-206EA 2190 DKK
HACH809-206
Electrode stand, E190
With its electrode arm that can move freely in all directions, this stand can hold up to four electrodes and a delivery tip. Easy to move from one sample to another within a circumference of 70 cm.