VWR® EZ-Vision®, PCR Ladder
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
N853-500ULEA 2270 DKK
N853-500UL
VWR® EZ-Vision®, PCR Ladder
Elektroforese reagenser Molekylærvægt markører
EZ-Vision® PCR DNA ladder is a pre-stained molecular weight ladder in loading buffer that allows immediate visualisation of DNA ladder post-electrophoresis. This non toxic, non mutagenic dye eliminates the need for ethidium bromide staining. Eight fragments range from 50 to 2000 bp with reference bands for easy identification.
  • 8 fragments ranging from 50 to 2000 bp
  • 'Load and go' formula with instant visualisation
  • Eliminates the use of ethidium bromide