Fluorescence cell analyser, Arthur™
Leverandør: NanoEnTek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Arthur™
734-2897EA 39470 DKK
734-2897
Fluorescence cell analyser, Arthur™
Celletæller Elektroniske celletællere
Image-based cell analyser with a three-channel (brightfield, green and red fluorescence) desktop image analyser, that allows to perform assays for cells in suspension including GFP and RFP expression, apoptosis, cell viability (live, dead and total cells), cell cycle and cell counting assays.
  • Accurate cell counting and analysis
  • Compatible with a wide range of eukaryotic cells
  • Rapid set-up and user friendly interface: No system maintenance required
Depending on the complexity of the assay, Arthur takes between 10 seconds and 2 minutes to count a typical assay with only 25 µl sample volume.