Anti-JUN Mouse Monoclonal Antibody [clone: 9F5]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF800592EA 5700 DKK
ORIGCF800592
Anti-JUN Mouse Monoclonal Antibody [clone: 9F5]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:500
Type: Primary
Antigen: JUN
Clonality: Monoclonal
Clone: 9F5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human